ContactUsLinkComponent
TICKOPUR R 36 - 1 LITRE, SPECIAL CLEANER, FOR THE ANALYTICAL APPLICATION, LASER TECHNOLOGY AND BLADE CLEANING, TENSIDE-FREE, CONCENT RATE, NON-FOAMING, GENTLE TO MATERIAL, DOSAGE 0.25 - 5 %, MILDLY ALKALINE, PH 9.9 (1 %) 1 * 1 L
Catalog # 142-1920
TICKOPUR R 36 - 1 LITRE, SPECIAL CLEANER, FOR THE ANALYTICAL APPLICATION, LASER TECHNOLOGY AND BLADE CLEANING, TENSIDE-FREE, CONCENT RATE, NON-FOAMING, GENTLE TO MATERIAL, DOSAGE 0.25 - 5 %, MILDLY ALKALINE, PH 9.9 (1 %) 1 * 1 L
Catalog # 142-1920
Frequently Bought Together