Cyclohexanone 99+% ACS
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99+% CAS No.: 108-94-1 Molecular Formula: C6H10O Formula Weight: 98.15 Flash Point: 47°(116°F) Physical Form: Liquid Density: 0.9478 Refractive Index: 1.4507 Melting Point: -26° Boiling Point: 154-156° Beilstein Reference: 385735 Merck Reference: 14,2726 UN No.: 1915 MDL No.: MFCD00001625
Synonyms: Pimelic Ketone
- CAS Number:108-94-1
- Formula:C₆H₁₀(=O)
- MDL Number:MFCD00001625
- Molecular Weight:98.14 g/mol
- Merck Index:14,2726
- Boiling Pt:154…156 °C
- Melting Pt:–26 °C
- Density:0.9478
- Flash Pt:47 °C (116 °F)
- Storage Temperature:Ambient
- EINECS:203-631-1
- UN:1915
- ADR:3,III
- REACH:01-2119453616-35
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 L



