Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
 

Cyclohexanone 99+% ACS

Glass bottle, 500 mL
Catalog # 33309.AP
Flammable
Irritant
Cyclohexanone 99+% ACS
Catalog # 33309.AP
CAS Number:  108-94-1
Cyclohexanone 99+% ACS
Cyclohexanone 99+% ACS

Glass bottle, 500 mL

CAS Number:  108-94-1
Price
1 530,00 Kč
Log in for availability
1 * 500 mL
Product Details & Documents

Purity: 99+% CAS No.: 108-94-1 Molecular Formula: C6H10O Formula Weight: 98.15 Flash Point: 47°(116°F) Physical Form: Liquid Density: 0.9478 Refractive Index: 1.4507 Melting Point: -26° Boiling Point: 154-156° Beilstein Reference: 385735 Merck Reference: 14,2726 UN No.: 1915 MDL No.: MFCD00001625

Synonyms:  Pimelic Ketone
  • CAS Number:108-94-1
  • Formula:C₆H₁₀(=O)
  • MDL Number:MFCD00001625
  • Molecular Weight:98.14 g/mol
  • Merck Index:14,2726
  • Boiling Pt:154…156 °C
  • Melting Pt:–26 °C
  • Density:0.9478
  • Flash Pt:47 °C (116 °F)
  • Storage Temperature:Ambient
  • EINECS:203-631-1
  • UN:1915
  • ADR:3,III
  • REACH:01-2119453616-35
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications