ContactUsLinkComponent
Combination pH electrode, no cable, S7 connector head, Diaphragm: Ceramic, Material: glass shaft, Electrolyte: KCl 3 mol/l refillabl e, Length: 400 mm, Diameter: 12 mm, Temperature range: -10 to 100 °C 1 * 1 items
Catalog # 620-4915
Supplier:  HAMILTON BONADUZ
Combination pH electrode, no cable, S7 connector head, Diaphragm: Ceramic, Material: glass shaft, Electrolyte: KCl 3 mol/l refillabl e, Length: 400 mm, Diameter: 12 mm, Temperature range: -10 to 100 °C 1 * 1 items
Catalog # 620-4915
Supplier:  HAMILTON BONADUZ
Frequently Bought Together