ContactUsLinkComponent
Container stainless steel with lid, maximum capacity 1L, maximum working capacity 500ml 1 * 1 items
Catalog # WARISS115
Supplier:  WARING BLENDOR
Container stainless steel with lid, maximum capacity 1L, maximum working capacity 500ml 1 * 1 items
Catalog # WARISS115
Supplier:  WARING BLENDOR
Frequently Bought Together