Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Retort stands, with M10 thread

Retort stands, with M10 thread

Retort stand with rubber feet and M10 thread, 210×130 mm, 1,6 kg
Katalogové číslo 241-0240
Dodavatel:   RSG
Retort stands, with M10 thread
Katalogové číslo 241-0240
Dodavatel:  RSG
Retort stands, with M10 thread
Retort stands, with M10 thread

Retort stand with rubber feet and M10 thread, 210×130 mm, 1,6 kg

Přejít na:
Price
3 220,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents

Retort stand bases made of stainless steel (Remanit 4301). With off-centre M10 thread.

Specifikace
  • Description:
    Retort stand with rubber feet and M10 thread
  • L×W:
    210×130 mm
  • Weight:
    1,6 kg