Jump to:
Product Details & Documents
- CAS Number:123-63-7
- Formula:C₆H₁₂O₃
- MDL Number:MFCD00036208
- Molecular Weight:132.16 g/mol
- Storage Temperature:Ambient
- ADR:3,III
- :M-Clarity™ quality level = MQ200
- EINECS:204-639-8
- UN:1264
Specifications
Specification Test Results
- Solubility:120 g/l (13 °C)
- Melting Point:12 °C
- Molar Mass:132.15 g/mol
- Boiling Point:124 °C (1013 hPa)
- Vapor Pressure:13.3 hPa (20 °C)
- Flash Point:27 °C
- Refractive Index:1.4049 (20 °C)
- Explosion Limit:1.3 - 17.0 %(V)
- Density:0.99 g/cm3 (20 °C)
- Ignition Point:235 °C
Specifications
- Pack type:Glass bottle
- Pk:100 ml



