KJELTABS CM 1 * 1.000 Tablet
Catalog # THOMAA50
Supplier: THOMPSON & CAPPER
Frequently Bought Together
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Test strips for free chlorine, MQuant®, Supelco®
Supplier: Merck
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach