Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
 

HPLC columns, LiChrospher®

Analytical, RP-select B, 60 Å, 5 µm, 3,00 mm, 250 mm
Katalogové číslo 1.50155.0001
Dodavatel:   Merck
HPLC columns, LiChrospher®
Katalogové číslo 1.50155.0001
Dodavatel:  Merck
HPLC columns, LiChrospher®
HPLC columns, LiChrospher®

Analytical, RP-select B, 60 Å, 5 µm, 3,00 mm, 250 mm

Přejít na:
Price
24 440,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents

LiChrospher® is Merck's reliable and versatile, traditionally-produced, spherical silica. LiChrospher® is available with different modifications.

LiChrospher® Si 60 and Si 100 are versatile HPLC spherical silica sorbents for normal phase chromatography.
LiChrospher® CN has polar and hydrophobic properties, so can be used as a less polar alternative to LiChrospher® Si 60 in normal phase applications, or as a less hydrophobic alternative to LiChrospher® RP-8 in reversed phase applications. In addition, selective charged interactions makes it even more versatile.
LiChrospher® NH₂ possesses polar and hydrophobic properties and can be used for normal phase, reversed phase and ion exchange chromatography. Typical applications are the separations of carbohydrates by reversed phase chromatography or the separation of nucleotides by ion exhange chromatography.

LiChrospher® DIOL has polar and hydrophobic properties, so can be used as a less polar alternative to LiChrospher® Si 60 in normal phase applications, or as a less hydrophobic alternative to LiChrospher® RP-8 in reversed phase applications. LiChrospher® DIOL is also suitable for exclusion chromatography.
LiChrospher® RP-select B is a highly reproducible, traditionally-produced spherical silica gel with reversed phase properties. This sorbent is ideal for the separation of basic compounds, but can also be used with neutral and acidic samples.

LiChrospher® RP-8, RP-18 and RP-18 endcapped are highly reproducible and versatile traditionally-produced spherical silica gel sorbents with reversed-phase properties. They are ideal for the separation of neutral, acidic and weak basic compounds.
LiChrospher® WP 300 RP-18 is a highly selective and reliable HPLC column for the separation of peptides and low molecular weight proteins. LiChrospher® WP 300 RP-18 enables the tailing-free separation of basic compounds, and is suitable for the separation of tRNA.
LiChrospher® PAH is a highly efficient and selective HPLC column, especially designed for the high resolution separation of 16 PAH (polycyclic aromatic hydrocarbons) according to EPA 610 and 550, plus benzo(e)pyrene and perylene.

LiChroCART® cartridges (2, 3, 4, 4,6 mm I-Ø) require the use of manu-CART® cartridge holder 1.51486.0001
LiChroCART® cartridges (10×250 mm) require the use of manu-CART® cartridge holder 1.51419.0001
Guard cartridges have to be connected to the LiChroCART® cartridges using manu-CART® cartridge holder 1.51486.0001.

LiChrospher® RP-8, RP-18 and RP-18 endcapped are highly reproducible and versatile traditionally produced spherical silica gel sorbents with reversed phase properties. They are well suited for the separation of neutral, acidic and weak basic compounds.

Specifikace
  • Column Format:
    Analytical
  • Ø int.:
    3,00 mm
  • Particle Size:
    5 µm
  • Pore size:
    60 Å
  • Length:
    250 mm
  • Seq:
    02
  • Phase:
    RP-select B
  • Cat. No.:
    1.50155.0001