ContactUsLinkComponent
6-Position Cell Holder platform with six (6) Single Cell Holders installed. Included with GENESYS 10Vis and GENESYS 10UV spectrophot 1 * 1 items
Catalog # SPET335927-000
6-Position Cell Holder platform with six (6) Single Cell Holders installed. Included with GENESYS 10Vis and GENESYS 10UV spectrophot 1 * 1 items
Catalog # SPET335927-000
Frequently Bought Together