Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
 

Visual colour kits, oxygen, MQuant®, Supelco®

Oxygen test, RP for 1.11107.0001, Winkler (modified), MTC, 100 Tests
Katalogové číslo 1.11152.0001
Dodavatel:   Merck
Visual colour kits, oxygen, MQuant®, Supelco®
Katalogové číslo 1.11152.0001
Dodavatel:  Merck
Číslo CAS:  
Visual colour kits, oxygen, MQuant®, Supelco®
Visual colour kits, oxygen, MQuant®, Supelco®

Oxygen test, RP for 1.11107.0001, Winkler (modified), MTC, 100 Tests

Číslo CAS:  
Price
4 910,00 Kč
Přihlásit se pro dostupnost
1 * 100 Tests
Product Details & Documents

Quick tests, visual colour comparison.

  • Results in approximately 2 minutes
  • Easy to use
  • Application-oriented measuring ranges
  • Portable
  • A range of economical refill packs (RP) are available

MQuant® titrimetric and colorimetric methods:

This range comprises both titrimetric and colorimetric tests for medium concentrations. With its broad range of available test parameters, these quick tests are the method of choice for aquaria, fish farming, aquatic ecology, environmental, cooling and boiler water analysis.

 : Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart and extensive instructions for use.

Specifikace

Specifikace

  • Chemical method:
    Winkler (modified)
  • Pk:
    100 Tests
  • Description:
    Oxygen test, RP for 1.11107.0001
  • Type:
    MTC
  • Cat. No.:
    1.11152.0001
  • M-Clarity quality level:
    MQ100

Specifikace

Accessories

Informace o produktu

  • Katalogové číslo
    Description
    Type
    Dostupnost
    Price
  • 1.14663.0001
    Oxygen reaction tube for 1.14662.0001 (MTC)
    MTC
    Přihlásit se pro dostupnost
    1 * 1 KS - 2 630,00 Kč