ContactUsLinkComponent
Magnetic stirrer TM96 for glass vessels up to maximale 500 ml, speed can be adjusted without discrete steps 0 - 1000 min-1, connecti on to TITRONIC® universal, TITRONIC® 97, TITRONIC® 96, TITRONIC® basic, TitroLine 96, TitroLine easy 1 * 1 items
Catalog # 661-4410
Supplier:  SI Analytics
Magnetic stirrer TM96 for glass vessels up to maximale 500 ml, speed can be adjusted without discrete steps 0 - 1000 min-1, connecti on to TITRONIC® universal, TITRONIC® 97, TITRONIC® 96, TITRONIC® basic, TitroLine 96, TitroLine easy 1 * 1 items
Catalog # 661-4410
Supplier:  SI Analytics
Frequently Bought Together