ContactUsLinkComponent
Cleaning solution L5104, DURAN® bottle, cleaning solution pepsin/hydrochloric acid 1 * 250 mL
Catalog # 662-6187
Supplier:  SI Analytics
Cleaning solution L5104, DURAN® bottle, cleaning solution pepsin/hydrochloric acid 1 * 250 mL
Catalog # 662-6187
Supplier:  SI Analytics