SPARE PARTS SET FOR PIPETTE KARTELL 100µL INCLUDING : 1 CONE + 2 PTFE SEAL + 2 O-RING + 1 CONE NUT 1 * 1 SET
KART1343
: KARTELL
SPARE PARTS SET FOR PIPETTE KARTELL 100µL INCLUDING : 1 CONE + 2 PTFE SEAL + 2 O-RING + 1 CONE NUT 1 * 1 SET
KART1343
: KARTELL
Frequently Bought Together
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection