Potassium acetate 99+% for analysis
Plastic bottle, 500 g
: Acetic acid potassium salt
- :127-08-2
- :CH₃COOK
- :MFCD00012458
- :98.14 g/mol
- :13,07687
- :292 °C
- :1,57 g/cm³ (25 °C)
- :>250 °C
- :Ambient
- :204-822-2
- Appearance:White Granular powder
- Infrared spectrum:Conforms
- Water:≤1.0 %
- pH:7.5 to 9.0 (5% soln. at 20°C)
- Titration with HClO4:99.0 to 101.0 %
- Chloride (Cl):≤50 ppm
- Sulfate (SO4):≤50 ppm
- Iron (Fe):≤20 ppm
- Heavy metals (as Pb):≤10 ppm
- Pack type:Plastic bottle
- Pk:500 g



