D-(-)-Fructose 99%
Plastic bottle, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
- CAS Number:57-48-7
- Formula:C₆H₁₂O₆
- MDL Number:MFCD00148910
- Molecular Weight:180.16 g/mol
- Merck Index:13,04295
- Boiling Pt:552 °C (1013 hPa)
- Melting Pt:103…105 °C (dec.)
- Density:1,59 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-333-3
Documents
Specifications
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Infrared spectrum:Conforms
- Assay:≥98.5 %
- Loss on drying:≤0.5 % (70°C, 4 hrs) (Vacuum)
- Sulfated ash:≤0.1 %
- Additional info:5-Hydroxymethylfurfural and rel. compounds: Passes
- Appearance of solution:Passes test
- Specific optical rotation:-91.0° to -93.5° (20°C, 589 nm) (c=10, H2O/NH3) (After 30 min, On dry substance)
- Chloride (Cl):≤50 ppm
- Sulfate (SO4):≤50 ppm
- Heavy metals (as Pb):≤10 ppm
- Foreign sugars:Passes test
Specifications
- Pack type:Plastic bottle
- Pk:500 g



