ContactUsLinkComponent
Alcoholometer, EUROPA classII, measuring range:90-100:0.1%vol, without thermometer, length:360mm, reference temperature: 20C, suita ble for government verification. 1 * 1 items
Catalog # AMARH848128
Supplier:  Amarell
Alcoholometer, EUROPA classII, measuring range:90-100:0.1%vol, without thermometer, length:360mm, reference temperature: 20C, suita ble for government verification. 1 * 1 items
Catalog # AMARH848128
Supplier:  Amarell
Frequently Bought Together