ContactUsLinkComponent
Alcoholometer, EUROPA classII, measuring range:80-90:0.1%vol, without thermometer, length:360mm, reference temperature: 20C, suitab le for government verification. 1 * 1 items
Catalog # AMARH848126
Supplier:  Amarell
Alcoholometer, EUROPA classII, measuring range:80-90:0.1%vol, without thermometer, length:360mm, reference temperature: 20C, suitab le for government verification. 1 * 1 items
Catalog # AMARH848126
Supplier:  Amarell
Frequently Bought Together