ContactUsLinkComponent
Areometer, DIN12791, L50 series, measuring range: 1.20-1.25:0.0005g/cm , without thermometer, reference temperature:20°C, length:335 mm, suitable for government verification. 1 * 1 items
Catalog # AMARH806224
Supplier:  Amarell
Areometer, DIN12791, L50 series, measuring range: 1.20-1.25:0.0005g/cm , without thermometer, reference temperature:20°C, length:335 mm, suitable for government verification. 1 * 1 items
Catalog # AMARH806224
Supplier:  Amarell
Frequently Bought Together