ContactUsLinkComponent
Areometer, DIN12791, L50 series, measuring range: 1.10-1.15:0.0005g/cm , without thermometer, reference temperature:20°C, length:335 mm, suitable for government verification. 1 * 1 items
Catalog # AMARH806220
Supplier:  Amarell
Areometer, DIN12791, L50 series, measuring range: 1.10-1.15:0.0005g/cm , without thermometer, reference temperature:20°C, length:335 mm, suitable for government verification. 1 * 1 items
Catalog # AMARH806220
Supplier:  Amarell
Frequently Bought Together