Areometer, DIN12791, L50 series, measuring range: 1-1.05:0.0005g/cm , without thermometer, reference temperature:20°C, length:335mm, suitable for government verification. 1 * 1 items
Catalog # AMARH806216
Supplier: Amarell
Frequently Bought Together