c-Src (B-12) mouse monoclonal IgG2a 200 µg/ml specific for an epitope mapping between amino acids 506-537 at the C-terminus of c- Src of human origin 1 * 0,1 mL
Catalog # SANTSC-8056
Supplier: Santa Cruz Biotechnology
Frequently Bought Together