Desiccator cabinets, SICCO Mini Basic series
Mini desiccator, Basic, (6,2 l), 3 chambers, 221×214×540 mm, 212×180×162 mm, 2,7 kg
467-0051
Clear, polycarbonate. Each compartment has one door with snap fit closing and integrated gasket ensuring easy opening and tight closure. Temperature resistant from −35 to +70 °C, suitable for use under normal pressure.
Robust construction, lightweight, portable and stackable
- Supplied with desiccant which is in a handy, resealable container
: Shelves must be ordered separately.
- Weight:2,7 kg
- W×D×H ext.:221×214×540 mm
- Description:Mini desiccator, Basic, (6,2 l)
- W×D×H int.:212×180×162 mm
- With:3 chambers



