ContactUsLinkComponent
Stirring bar, magnetic, elliptical, egg shaped, PTFE, for 12000 and 22000ml round bottomed flask, size 76 x 19.1mm. 1 * 1 items
Catalog # BELAF371300003
Stirring bar, magnetic, elliptical, egg shaped, PTFE, for 12000 and 22000ml round bottomed flask, size 76 x 19.1mm. 1 * 1 items
Catalog # BELAF371300003
Frequently Bought Together