ContactUsLinkComponent
Stirring bar, micro (flea), magnetic, PTFE, white, size 7x2mm, square ends, small sizes for stirring small volumes in vessels such a s vials, tubes, and gradient makers. 1 * 1 items
Catalog # BELAF371190007
Stirring bar, micro (flea), magnetic, PTFE, white, size 7x2mm, square ends, small sizes for stirring small volumes in vessels such a s vials, tubes, and gradient makers. 1 * 1 items
Catalog # BELAF371190007
Frequently Bought Together