Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
 

Micro scissors

Straight, sharp/sharp, cutting length 15 mm, 105 mm
  233-1101
Micro scissors
  233-1101
Micro scissors
Micro scissors

Straight, sharp/sharp, cutting length 15 mm, 105 mm

 
10 920,00 Kč
   
1 * 1 KS
 

Stainless steel.

 
  • Type:
    Straight, sharp/sharp, cutting length 15 mm
  • Length:
    105 mm
  • Seq:
    11