ContactUsLinkComponent
Syringe,10ul,fixed needle,51 x 0.52mm, point style 2.Hamilton. 1 * 1 items
Catalog # 549-1112
Supplier:  HAMILTON BONADUZ
Syringe,10ul,fixed needle,51 x 0.52mm, point style 2.Hamilton. 1 * 1 items
Catalog # 549-1112
Supplier:  HAMILTON BONADUZ
Frequently Bought Together