ANION STANDARD NITRATE 1000PPM 1 * 100 mL
REGEICAS04
: REAGECON
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Water, GPR RECTAPUR®
: VWR Chemicals
Diethyl ether, anhydrous (max. 0.005% H₂O) ≥99.8% stabilised
: VWR Chemicals
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
: VWR Chemicals
Ethyl acetate ≥99.7%, PESTINORM® for pesticide residue analysis
: VWR Chemicals
m-Xylene, extra dry 99%, AcroSeal®
: Thermo Fisher Scientific
Titrants and reagents for volumetric Karl Fischer titrations, HYDRANAL™, Fluka™
: Honeywell Chemicals