ANION STANDARD CHLORIDE 1000PPM 1 * 100 mL
REGEICAS02
: REAGECON
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Volumetric Flasks, Class A, Glass, Clear, NS Plastic Stopper
: VWR Collection
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
Special pipette tips, Finntip™
: Thermo Fisher Scientific
VWR®, Nitrile Gloves
: VWR Collection