BUFFER SOLUTION PH3,5 1 * 1 L
Catalog # REGE4000600
Supplier: REAGECON
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® Bulb Pipettes, Two Marks, Class AS
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
pH buffers, ready to use
Supplier: WTW
Ammonium formate, (max. 3% H₂O) 97%
Supplier: Thermo Fisher Scientific
Chromotropic acid disodium salt dihydrate ACS
Supplier: Thermo Fisher Scientific