Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
 

Bromomethyl methyl ether

Bottle
Catalog # APOSOR11138-2.5KG
Flammable
Irritant
Health
New Product
Supplier:   Apollo Scientific
Bromomethyl methyl ether
Catalog # APOSOR11138-2.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  13057-17-5
Bromomethyl methyl ether

Bottle

CAS Number:  13057-17-5
Price
53 820,00 Kč
Log in for availability
1 * 2,5 kg
Product Details & Documents
  • CAS Number:13057-17-5
  • Formula:C₂H₅BrO
  • MDL Number:MFCD00000171
  • Molecular Weight:124.97 g/mol
  • Boiling Pt:27 °C
  • Melting Pt:87 °C
  • Density:1.531 g/cm³ (25 °C)
  • Storage Temperature:Refrigerator
  • UN:1992
  • ADR:3+6.1,III
Specifications

Specifications

  • Pack type:
    Bottle

Specifications