Bromomethyl methyl ether
Bottle
Katalogové číslo APOSOR11138-1G
New Product
Dodavatel: Apollo Scientific
Bromomethyl methyl ether
Katalogové číslo APOSOR11138-1G
Dodavatel: Apollo Scientific
New Product
Číslo CAS: 13057-17-5
Přejít na:
Product Details & Documents
- Číslo CAS:13057-17-5
- Vzorec:C₂H₅BrO
- MDL číslo:MFCD00000171
- Molecular Weight:124.97 g/mol
- Bod varu:27 °C
- Bod tání:87 °C
- Hustota:1.531 g/cm³ (25 °C)
- Teplota pro skladování:Refrigerator
- UN:1992
- ADR:3+6.1,III
Specifikace
Specifikace
- Pack type:Bottle
Specifikace
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



