Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
 

EBPC ≥95%

Bottle
Catalog # APOSBUP08358-100MG
New Product
Supplier:   Apollo Scientific
EBPC ≥95%
Catalog # APOSBUP08358-100MG
Supplier:  Apollo Scientific
New Product
CAS Number:  4450-98-0
EBPC ≥95%

Bottle

CAS Number:  4450-98-0
Price
21 280,00 Kč
Log in for availability
1 * 100 mg
Product Details & Documents

Usage: Biochemical/life science applications.

  • CAS Number:4450-98-0
  • Formula:C₁₄H₁₅NO₄
  • Molecular Weight:261,27 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications