N,O-Di-boc-hydroxylamine
Bottle
APOSOR46242-500G
New Product
: N,O-Di-Boc-hydroxylamine, tert-Butyl N-(tert-Butoxycarbonyloxy)carbamate, N,O-Bis(tert-butoxycarbonyl)hydroxylamine, tert-Butyl (tert-butoxycarbonyl)oxycarbamate, N-(tert-Butoxycarbonyloxy)carbamic acid tert-Butyl ester
- :85006-25-3
- :C₁₀H₁₉NO₅
- :MFCD00034797
- :233.264 g/mol
- :67…70 °C
- :Freezer
- :285-055-0
- Pack type:Bottle