Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Pitstop® 2-100 - negative control

Pitstop® 2-100 - negative control

Catalog # ABCAAB144658-5
Supplier:   Abcam
Pitstop® 2-100 - negative control
Catalog # ABCAAB144658-5
Supplier:  Abcam
CAS Number:  
Pitstop® 2-100 - negative control
CAS Number:  
Price
7 350,00 Kč
Log in for availability
1 * 5 mg
Product Details & Documents

Negative control for clathrin inhibitor Pitstop® 2-100. Structurally related to Pitstop® 2-100 but unable to associate with the clathrin terminal domain. No effect on clathrin-mediated endocytosis (CME) of transferrin or major histocompatibility complex 1 (MHCI) uptake.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  Pitstop™ 2-100 - negative control
  • Molecular Weight:463,39 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% Pitstop™ 2-100 - negative control
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.