Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
 

3,3',5,5'-Tetramethylbenzidine free base

Supplier:   G-Biosciences
3,3',5,5'-Tetramethylbenzidine free base
Supplier:  G-Biosciences
Certificates
3,3',5,5'-Tetramethylbenzidine free base

2 Options Available Below

Product Details & Documents

TMB is a widely used chromogenic substrate for horseradish peroxidase (HRP) in colorimetric assays. As a free base, it is typically dissolved in an organic solvent or buffer to prepare TMB substrate solutions for ELISA, Western blotting, and immunohistochemistry.

  • Sensitive HRP Substrate: Enables high-sensitivity detection in enzyme-linked assays
  • Colorimetric Detection: Produces a strong blue signal, with a yellow endpoint upon acid stop for dual-wavelength measurement
  • Flexible Formulation: Free base form allows custom preparation for specific assay conditions
  • Versatile Applications: Ideal for ELISA, Western blotting, and immunoassays in diagnostics and research
  • Low Background: Provides high signal-to-noise ratio for accurate quantification
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 23 740,00 Kč
    23 740,00 Kč
    1 * 50 g
       23 740,00 Kč

  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 37 990,00 Kč
    37 990,00 Kč
    1 * 100 g
       37 990,00 Kč