G-Biosciences S.O.C Medium for Bacterial Transformation
S.O.C Medium is used in bacterial cell transformation after heat-shock treatment to the DNA-competent cell mixture to obtain maximal transformation efficiency.
Sterile filtered and quality checked
Composition: 2% Tryptone, 0.5% Yeast extract, 10 mM NaCl, 2.5 mM KCl, 10 mM MgCl₂, 10 mM MgSO₄ and 20 mM glucose.
pH of the S.O.C Medium: 6,8 ±0,2.
Frequently Bought Together