ContactUsLinkComponent
VWR® DOSATEST® Test Strips, Nitrite/pH
VWR® DOSATEST® Test Strips, Nitrite/pH
Catalog # 92533.601
Supplier:  VWR Chemicals
New Product
VWR® DOSATEST® Test Strips, Nitrite/pH
Catalog # 92533.601
Supplier:  VWR Chemicals
New Product
Specifications
  • Description:
    Multi parameter semi-quantitative test strips, 100 tests
Specifications
About this item
Dosatest® pH/Nitrite test strip enables quick and reliable determination of the pH value and nitrite content in water–based liquids.
  • Range: Nitrite - 0, 1, 5, 10, 20, 40 and 80  mg/L NO₂⁻, pH - 6.0, 6.4, 6.7, 7., 7.3, 7.6, 7.9, 8.2, 8.4, 8.6, 8.8, 9.0, 9.3 and 9.6
    Shelf life: 2.5 Year(s)
    Storage temperature: 4 to 30 °C (39 to 86 °F)
    Size: 100 test strips in a tube, instruction leaflet
The easy handling makes the test strip particularly user–friendly: simply dip it into the sample, wait and read off the result - in 60 seconds. DOSATEST® pH/Nitrite is therefore ideal for the analysis of water–based coolants. Regular monitoring of the cooling lubricant emulsion is not only a decisive prerequisite for its optimal use, but also important for the safety of the user. Low pH levels reduce the corrosion protection and stability of the emulsion. High pH and nitrite levels can pose a health risk to the operator.