ContactUsLinkComponent
SP Bel-Art Switch-Grid Test Tube Rack, Complete Set, Interchangeable Grids Included
SP Bel-Art Switch-Grid Test Tube Rack, Complete Set, Interchangeable Grids Included
Catalog # BELAF18745-1000
New Product
SP Bel-Art Switch-Grid Test Tube Rack, Complete Set, Interchangeable Grids Included
Catalog # BELAF18745-1000
New Product
Specifications
  • For tubes:
    10 - 30 mm
  • No. of Holes:
    12, 24, 40, 60, 84
  • Material:
    Polyoxymethylene
  • W×D×H:
    318×111×670 mm
Specifications
About this item
The innovative Switch-Grid rack utilises interchangeable grids for ultimate flexibility. This split-frame rack holds up to two different tube sizes at the same time and allows users to quickly switch grids to hold other sizes.
  • 10 to 30 mm diameter test tubes fit in 5 color-coded grid inserts 
  • Can be easily cleaned with common lab detergents, ethanol, or isopropyl alcohol
  • Three-tier design securely holds tubes in place while non-slip rubber feet prevent sliding on the lab bench
  • Rack available without grids for use with spare grids
This rack is a smart solution for test tube organization and streamlining the lab's workflow. Constructed from chemical-resistant polyoxymethylene plastic, both the frame and grids are autoclavable at 121 °C.