ContactUsLinkComponent
Washing Bottle Lower Part for 3410* and 3425* Standard Joint NSK 45/40
Washing Bottle Lower Part for 3410* and 3425* Standard Joint NSK 45/40
Catalog # ROBU34270
Supplier:  ROBU GLASFILTER
Washing Bottle Lower Part for 3410* and 3425* Standard Joint NSK 45/40
Catalog # ROBU34270
Supplier:  ROBU GLASFILTER
Specifications
  • NS:
    45/40
  • Capacity:
    250 ml
  • Description:
    Washing bottle lower part
  • Pk:
    1
  • For bottles:
    Fits 3410* and 3425*
Specifications
About this item
ROBU® washing bottle lower part, made from durable borosilicate glass, serves as the liquid reservoir in gas absorption processes.
  • High-quality borosilicate glass
  • High chemical and thermal resistance
  • Compatible with standard ground joint heads
  • Robust and reusable
  • Ideal for use with VitraPOR® heads
  • Clear visibility of contents
  • NSK 45/40
The lower part of the ROBU washing bottle is crafted from high-quality borosilicate glass, ensuring exceptional resistance to chemicals and thermal stress. Designed to serve as the receiving vessel in gas absorption systems, it securely holds liquids while allowing visual monitoring of the absorption process. Compatible with ROBU heads with standard joints and VitraPOR filter discs, this lower part forms a reliable, durable solution for efficient gas scrubbing applications in laboratories and industrial settings.
Certifications:
In compliance with ISO 4796-80 and ASTM E128-99 standards.