ContactUsLinkComponent
Proteometer-L Full Kit Reagent (1000)
Proteometer-L Full Kit Reagent (1000)
Catalog # NOVIKIT-PL-007-01B
Supplier:  NOVILYTIC
New Product
CAS Number:  
undefined
Proteometer-L Full Kit Reagent (1000)
Catalog # NOVIKIT-PL-007-01B
Supplier:  NOVILYTIC
New Product
CAS Number:  
Specifications
  • Description:
    Proteometer-L full kit reagent (1000)
Specifications
About this item
Fluorescent reagent for the Proteometer-L full kit. This is sold separately due to shipping conditions. Store at −20 °C.
  • One assay for two critical attributes - titer + aggregate measurement
  • No protein A purification required: Reduces costs
  • No sample pretreatment: Reduces hours of sample prep
  • No method development: Reduces time until method usage
  • Direct analysis from CFB: HCP removal not needed
  • Plug-and-play for any LC with fluorescence detector: Easy adoption, no software needed
  • Low sample volume: Reduces impact on processing
Crimped, sealed, ready for use with the Proteometer-L full kit (NOVIKIT-PL-007-01A) which is purchased separately. This kit is reagent only and pairs with our Proteometer-L full kit.
Reagent to be stored at −20 °C.
Ordering information: Please add Proteometer-L full kit seperately (NOVIKIT-PL-007-01A).
Delivery:
Includes 14× Proteometer-L fluorescent reagent vials - allows for analysis up to 1000 samples.
Packaging: Packaged separately from the Proteometer-UFT full kit due to storage conditions.