Proteometer-L full kit part 1/2 (1000 titer and aggregate assays)
Catalog # NOVIKIT-PL-007-01A
Supplier: NOVILYTIC
New Product
Specifications
- Description:Proteometer-L full kit (1000), for use with any LC equipped with a fluorescence detector
Specifications
About this item
1000 Titer + aggregate assays. Use with any LC. For human mAbs, multi-specifics, recombinant proteins.
- One assay for two critical attributes - titer + aggregate measurement
- No protein A purification required: Reduces costs
- No sample pretreatment: Reduces hours of sample prep
- No method development: Reduces time until method usage
- Direct analysis from CFB: HCP removal not needed
- Plug-and-play for any LC with fluorescence detector: Easy adoption, no software needed
- Low sample volume: Reduces impact on processing
Eliminate sample prep and method development. The kit includes the Proteometer reactor, reconstitution reagent, buffers, and other parts needed for a 'plug-and-play' use with any liquid chromatograph that has a fluoresence detector.
Accessories information: See the Proteometer-L reactor, Proteometer-L buffer packets, and Proteometer-L reagent series for accessory component replacement options.
Ordering information: Glassware not included. Please add Proteometer-L full kit fluorescent reagent seperately (NOVIKIT-PL-007-01B).
Delivery:
Includes 1× Proteometer-L reactor, 14× Proteometer-L reagent vials, 14× Proteometer-L buffer packets, 1× reconstitution reagent. Allows for analysis up to 1000 samples. Please add Proteometer-L full kit fluorescent reagent seperately (NOVIKIT-PL-007-01B).
Packaging: Supplied in a custom box with instructions and how-to video. No refrigeration or special handling required.