ContactUsLinkComponent
HCL-Absorber
HCL-Absorber
  ROBU18901
undefined
HCL-Absorber
  ROBU18901
  • Description:
    HCL-absorber, Ø×H 25×190 mm
ROBU® HCL-Absorber efficiently captures and absorbs hydrogen chloride gas (HCl) in laboratory or industrial applications. Made of high-quality materials, it provides reliable protection against harmful fumes while offering durability and ease of use.
  • Efficient HCl gas absorption
  • High chemical resistance
  • Reliable protection against harmful fume
  • Durable and long-lasting
  • Ideal for laboratory and industrial use
  • Easy to integrate into existing systems
  • Reusable and autoclavable
ROBU HCL-Absorber is designed to effectively capture and neutralize hydrogen chloride (HCl) gas, making it ideal for use in laboratory environments and industrial applications where HCl fumes are generated. Constructed from durable materials with high chemical resistance, this absorber ensures reliable protection against harmful gas exposure.
It is easy to incorporate into existing filtration and ventilation systems, offering a cost-effective solution for maintaining safe working conditions. The HCL-Absorber is reusable, autoclavable, and designed to withstand demanding environments, ensuring long-term performance and minimal maintenance.
Standard ground joint: NSH 29/32 | NSK 29/32
Material: Borosilicate glass 3.3
 :
In compliance with ISO 4796-80 and ASTM E128-99 standards.