ContactUsLinkComponent
Cap with Standard Joint NSH 29/32
Cap with Standard Joint NSH 29/32
Catalog # ROBU37370
Supplier:  ROBU GLASFILTER
undefined
Cap with Standard Joint NSH 29/32
Catalog # ROBU37370
Supplier:  ROBU GLASFILTER
Specifications
  • Cone size:
    NSK 29/32
  • Description:
    Cap with standard joint
  • Pk:
    1
  • For bottles:
    Fits 3735*
Specifications
About this item
ROBU® cap with a standard joint provides secure sealing and reliable connection for various industrial applications. Ideal for protecting contents and ensuring efficient operation. If the product includes a filter, the VitraPOR® brand filter offers additional filtration capabilities.
  • Reliable sealing
  • Prevents leaks or contamination
  • Standard joint compatibility
  • Fits with standard joint systems NSH 29/32
  • Durable material
  • Versatile use
This ROBU cap with a standard joint is designed to offer excellent performance in sealing and protection, especially in industries where reliability and durability are crucial. Made from high-quality materials, it ensures a long-lasting and secure fit, preventing the escape of contents and maintaining system integrity. Its compatibility with standard joint systems makes it a versatile option for laboratories, manufacturing plants, and other technical applications, where quick and efficient sealing is needed. If the product is equipped with a filter, the VitraPOR brand filter provides superior filtration, making it an ideal solution for processes requiring precision and cleanliness.