Order Entry
Czech Republic
ContactUsLinkComponent
Soft-Zellin® C Alcohol Swabs
Soft-Zellin® C Alcohol Swabs
Catalog # HART288887
Supplier:  HARTMANN PAUL
New Product
Soft-Zellin® C Alcohol Swabs
Catalog # HART288887
Supplier:  HARTMANN PAUL
New Product

Specifications

  • Description:
    Alcohol swab, 60×30 mm, folded to 30×30 mm

Specifications

About this item

Soft-Zellin C is an alcohol swab made of nonwoven fabric, impregnated with isopropyl alcohol (70% v/v).

  • Ready to use

    Flexible and soft

    Suitable for cleansing the skin

Material nonvowen: 40% rayon, 30%, polyester 30% ES fiber.

Certifications:

Product is classified as cosmetic according to EC 1223/2009.

Packaging:

Individually sealed and packed in folding boxes (100 pieces) and then into transport cartons. Material Packed aluminum compound foil.

Caution:

Not suitable for children under one year of age.