ContactUsLinkComponent
Proteometer-L Starter Kit (200 Titer)
Proteometer-L Starter Kit (200 Titer)
Catalog # NOVIKIT-PL-002-01A
Supplier:  NOVILYTIC
New Product
undefined
Proteometer-L Starter Kit (200 Titer)
Catalog # NOVIKIT-PL-002-01A
Supplier:  NOVILYTIC
New Product
Specifications
  • Description:
    Proteometer-L starter kit (200), for use with any LC equipped with a fluorescence detector
Specifications
About this item
200 Titer + aggregate assays (~70/day). Use with any LC. For human mAbs, multi-specifics, recombinant proteins.
  • mAb titer + aggregate measurement: one assay for two critical attributes
  • No Protein A purification required: Reduces costs
  • No sample pretreatment: Reduces hours of sample prep
  • No method development: Reduces time until method useage
  • Direct analysis from CFB: HCP removal not needed
  • Plug-and-play for any LC with fluorescence detector: Easy adoption, no software needed
  • Low sample volume: reduces impact on processing
Eliminate sample prep and method development. The kit includes the Proteometer Reactor, reconstitution reagent, buffers, and other parts needed for 'plug-and-play' use with any liquid chromatograph that has a fluoresence detector.
Accessories information: See the Proteometer-L reactor, Proteometer-L buffer packets, and Proteometer-L reagent series for accessory component replacement options.
Ordering information: Glassware not included. Please add Proteometer-L fluorescent reagent seperately (NOVIKIT-PL-002-01B).
Delivery: Includes 1× Proteometer-L reactor, 4× Proteometer-L reagent vials, 4× Proteometer-L buffer packets, 1× reconstitution reagent - allows for analysis up to 200 samples.
Packaging: Supplied in a custom box with instructions and how-to video. No refrigeration or special handling required.
Reagent component
Product Information