Order Entry
Czech Republic
ContactUsLinkComponent
TOYOPEARL® Super A Affinity Chromatography Protein Resins
TOYOPEARL® Super A Affinity Chromatography Protein Resins
Supplier:  TOSOH Bioscience
New Product
Certificates
TOYOPEARL® Super A Affinity Chromatography Protein Resins
Supplier:  TOSOH Bioscience
New Product

Specifications

  • pH range:
    3 - 12 (operational); 2 - 14 (CIP)
  • Matrix:
    Hydroxylated methacrylic polymer
  • Ligand:
    Modified, code-optimised C domain, hexameric ligand with multipoint attachment
  • Binding capacity:
    72 mg mAb/ml resin @ 5 min RT; 60 mg mAb/ml resin @ 3 min RT
  • Concentration:
    Endotoxin concentration: Max 10 EU/ml
  • Flow rate:
    10 - 600 cm/h (linear)
  • Formulation:
    72% slurry in 20% ethanol

Specifications

Frequently Bought Together