ContactUsLinkComponent
Cubis® II Manual Mass Comparators
New Product
undefined
Cubis® II Manual Mass Comparators
New Product
  • Catalog No:
  • SARBMCA106MC
  • SARBMCA206MC
  • SARBMCA1004MC
  • SARBMCA1005MC
  • SARBMCA605MC
  • SARBMCA66MC
  • SARBMCA36MC
  • Operating temperature range:
  • 5…40 °C
  • 5…40 °C
  • 5…40 °C
  • 5…40 °C
  • 5…40 °C
  • 5…40 °C
  • 5…40 °C
  • Plug type:
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Communication interface:
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • Voltage:
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
  • 100 - 240 V
Frequently Bought Together