VWR® Autoclavable Biohazard Bags, Polypropylene, Yellow
Supplier: VWR Collection
Certificates
About this item
Sturdy, temperature resistant bags, ideal for disposing of non hazardous waste or hazardous waste that must first be autoclaved.
Autoclavable up to 135 °C (275 °F) maximum
- 20 cm twist ties included allows bag to expand and vent steam during cooling
- Bags meet the requirements of the 165 g puncture inspection standard as described in ASTM D1709
- Bag is printed with the universal biohazard symbol
- Bag has temperature indicator, text 'Contents Have Been Autoclaved' turns dark to indicates successful autoclaving
- Highly resistant to tears and punctures
Can also be used for autoclaving other products. Supplied complete with wire closures that allow expansion of the bag and ventilation when cooling.
Certifications:
Certifications: ISO 9001:2015, Sedex.