Culture Media, Ready To Use, In Plastic Tubes
Dodavatel: LIOFILCHEM
New Product
Certifikáty
O tomto produktu
Microbiology culture media in plastic tubes are utilised for cultivating, storing and observing microorganisms in various microbiological culture applications. Plastic tubes are increasingly being used for culture media instead of glass tubes, offering advantages compared to traditional glass tubes.
Reduced weight and lower CO₂ emissions: Plastic tubes are lighter than glass, resulting in lower transportation costs and a reduced carbon footprint
- Enhanced safety: Plastic tubes are less likely to break during handling and transport, improving laboratory safety
- Shelf life: Two years for most of the products (One year for the others)
Frequently Bought Together